Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl cis-13-docosenoate ≥90.0%

Supplier: Decon Laboratories Ltd

D1017-500ML D1017-25ML
TCD1017-500MLEA 938 USD InStock
TCD1017-500ML TCD1017-025ML
Methyl cis-13-docosenoate ≥90.0%
Methyl cis-13-docosenoate
CAS Number: 1120-34-9
MDL Number: MFCD00027343
Molecular Formula: C23H44O2
Molecular Weight: 352.60
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 222
Flash Point (°C): 33
Specific Gravity (20/20): 0.87

Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: cis-13-Docosenoic Acid Methyl Ester, Erucic Acid Methyl Ester, Methyl Erucate


Formula: C₂₃H₄₄O₂
Boiling Pt: 222 °C
Density: 0.87 (20 °C)
Flash Pt: 33 °C
MDL Number: MFCD00027343
CAS Number: 1120-34-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1909a53bee9047966e687b3889939bf6ec8e04a8&vid=1909a53bee9047966e687b3889939bf6ec8e04a8&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >90.0% (GC)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty