D1023-1G
D1023-100MG
TCD1023-1GEA
133
USD
InStock
TCD1023-1G
TCD1023-100MG
Nε-(2,4-dinitrophenyl)-L-lysine hydrochloride ≥98.0% (by HPLC)
Nepsilon-(2,4-dinitrophenyl)-L-lysine hydrochloride
CAS Number: 14401-10-6
MDL Number: MFCD00036383
Molecular Formula: C12H16N4O6
Molecular Weight: 348.74
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 19 deg (C=2, MeOH)
Product Categories: Bioscience, Side Chain Protected Amino Acids (N,C-termini Free)
Other Product Names / Synonyms: Nepsilon-Dnp-L-lysine Hydrochloride, H-Lys(Dnp)-OH.HCl