Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2,4-Dinitrophenyl)-L-phenylalanine ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd

D1035-100MG D1035-1G
TCD1035-100MGEA 34 USD InStock
TCD1035-100MG TCD1035-1G
N-(2,4-Dinitrophenyl)-L-phenylalanine ≥98.0% (by HPLC, titration analysis)
N-(2,4-Dinitrophenyl)-L-phenylalanine
CAS Number: 1655-54-5
MDL Number: MFCD00038099
Molecular Formula: C15H13N3O6
Molecular Weight: 331.28
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 187
Specific rotation [a]20/D: -94.5 deg (C=1, AcOH)

Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Dnp-L-phenylalanine, Dnp-Phe-OH


Formula: C₁₅H₁₃N₃O₆
Melting Pt: 187 °C
MDL Number: MFCD00038099
CAS Number: 1655-54-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c62cd90245cc10f8d5ffd068d18a1aec6c4be1d&vid=7c62cd90245cc10f8d5ffd068d18a1aec6c4be1d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty