Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-2,3-Dibromopropionic acid ≥97.0% for synthesis

Supplier: Decon Laboratories Ltd

D1383-25G D1383-500G
TCD1383-025GEA 23 USD InStock
TCD1383-025G TCD1383-500G
(±)-2,3-Dibromopropionic acid ≥97.0% for synthesis
(±)-2,3-Dibromopropionic acid
CAS Number: 600-05-5
MDL Number: MFCD00004212
Molecular Formula: C3H4Br2O2
Molecular Weight: 231.87
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Boiling point (°C): 119
Melting point (°C): 65
Storage Temperature: 0-10°C


Formula: BrH₂CBrCHCOOH
Boiling Pt: 119 °C
Melting Pt: 65 °C
MDL Number: MFCD00004212
CAS Number: 600-05-5
EINECS: 209-981-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e84fcb45e57a926c234c749aa8b9f2a6ed2c5483&vid=e84fcb45e57a926c234c749aa8b9f2a6ed2c5483&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty