Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dihydro-2H-pyrido[1,2-a]pyrimidin-2-one ≥98.0%

Supplier: Decon Laboratories Ltd

D1393-5G D1393-25G
TCD1393-005GEA 53 USD InStock
TCD1393-005G TCD1393-025G
3,4-Dihydro-2H-pyrido[1,2-a]pyrimidin-2-one ≥98.0%
3,4-Dihydro-2H-pyrido[1,2-a]pyrimidin-2-one
CAS Number: 5439-14-5
MDL Number: MFCD00042798
Molecular Formula: C8H8N2O
Molecular Weight: 148.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification
Other Product Names / Synonyms: Acid Captor H


Formula: C₈H₈N₂O
MW: 148.17 g/mol
MDL Number: MFCD00042798
CAS Number: 5439-14-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2e8e9f51fd2ad1584140d7e825fe3227392e523&vid=b2e8e9f51fd2ad1584140d7e825fe3227392e523&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty