Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Dimethyltetrahydrothiazole ≥98.0%

Supplier: Decon Laboratories Ltd

D1491-5ML D1491-25ML
TCD1491-005MLEA 31 USD InStock
TCD1491-005ML TCD1491-25ML
2,2-Dimethyltetrahydrothiazole ≥98.0%
2,2-Dimethyltetrahydrothiazole
CAS Number: 19351-18-9
MDL Number: MFCD00014488
Molecular Formula: C5H11NS
Molecular Weight: 117.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 61
Flash Point (°C): 55
Specific Gravity (20/20): 1.02


Formula: C₅H₁₁NS
Boiling Pt: 61 °C
Density: 1.02 (20 °C)
Flash Pt: 55 °C
MDL Number: MFCD00014488
CAS Number: 19351-18-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3989a80c7bffa7655438e2912ab080d677655e4b&vid=3989a80c7bffa7655438e2912ab080d677655e4b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty