Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dimethylhexadecylamine ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

D1568-500ML D1568-25ML
TCD1568-500MLEA 127 USD InStock
TCD1568-500ML TCD1568-025ML
N,N-Dimethylhexadecylamine ≥98.0% (by titrimetric analysis)
N,N-Dimethylhexadecylamine
CAS Number: 112-69-6
MDL Number: MFCD00015086
Molecular Formula: C18H39N
Molecular Weight: 269.52
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 148
Flash Point (°C): 147
Specific Gravity (20/20): 0.80

Other Product Names / Synonyms: 1-(Dimethylamino)hexadecane, N,N-Dimethylcetylamine, N,N-Dimethylpalmitylamine


Formula: C₁₈H₃₉N
Boiling Pt: 148 °C
Density: 0.80 (20 °C)
Flash Pt: 147 °C
MDL Number: MFCD00015086
CAS Number: 112-69-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58f579bc63280b4c4e7027d0f0a7193e7fecc01d&vid=58f579bc63280b4c4e7027d0f0a7193e7fecc01d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty