Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Diethylamino)propionic acid hydrochloride ≥98.0%

Supplier: Decon Laboratories Ltd

D1713-25G D1713-5G
TCD1713-25GEA 389 USD InStock
TCD1713-25G TCD1713-005G
3-(Diethylamino)propionic acid hydrochloride ≥98.0%
3-(Diethylamino)propionic acid hydrochloride
CAS Number: 15674-67-6
MDL Number: MFCD00012611
Molecular Formula: C7H15NO2
Molecular Weight: 181.66
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 146

Other Product Names / Synonyms: N,N-Diethyl-beta-alanine Hydrochloride


Formula: C₇H₁₅NO₂
MW: 181.66 g/mol
Melting Pt: 146 °C
MDL Number: MFCD00012611
CAS Number: 15674-67-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dff69d80b3cdf5633c2f66d7cdb61acf813e0275&vid=dff69d80b3cdf5633c2f66d7cdb61acf813e0275&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty