D1733-25ML
D1733-500ML
TCD1733-025MLEA
20
USD
InStock
TCD1733-025ML
TCD1733-500ML
N,N,2,2-Tetramethyl-1,3-propanediamine ≥98.0%
N,N,2,2-Tetramethyl-1,3-propanediamine
CAS Number: 53369-71-4
MDL Number: MFCD00044018
Molecular Formula: C7H18N2
Molecular Weight: 130.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 153
Flash Point (°C): 35
Specific Gravity (20/20): 0.82
Other Product Names / Synonyms: N,N-Dimethylneopentanediamine