Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Diphenylpropionic acid ≥98.0%

Supplier: Decon Laboratories Ltd

D1790-25G D1790-5G
TCD1790-025GEA 485 USD InStock
TCD1790-025G TCD1790-005G
2,3-Diphenylpropionic acid ≥98.0%
2,3-Diphenylpropionic acid
CAS Number: 3333-15-1 MDL Number: MFCD00044405 Molecular Formula: C15H14O2 Molecular Weight: 226.28 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Boiling point (°C): 340 Melting point (°C): 89


Formula: C₁₅H₁₄O₂
Boiling Pt: 340 °C
Melting Pt: 89 °C
MDL Number: MFCD00044405
CAS Number: 3333-15-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ffb3a1527dc2053107dd6de26c0f52c404ba487&vid=9ffb3a1527dc2053107dd6de26c0f52c404ba487&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty