Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(-)-N-(3,5-Dinitrobenzoyl)-α-phenylglycine ≥98.0% for synthesis

Supplier: Decon Laboratories Ltd

D1853-1G D1853-5G
TCD1853-1GEA 38.7 USD InStock
TCD1853-1G TCD1853-005G
(R)-(-)-N-(3,5-Dinitrobenzoyl)-α-phenylglycine ≥98.0% for synthesis
(R)-(-)-N-(3,5-Dinitrobenzoyl)-α-phenylglycine
CAS Number: 74927-72-3
MDL Number: MFCD00010134
Molecular Formula: C15H11N3O7
Molecular Weight: 345.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -102 deg (C=1, THF)


Formula: C₁₅H₁₁N₃O₇
MW: 345.26 g/mol
MDL Number: MFCD00010134
CAS Number: 74927-72-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fee2c381e71213d31a7bfca9f06f9dd51be4c59d&vid=fee2c381e71213d31a7bfca9f06f9dd51be4c59d&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty