Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,6-Diaminocarbazole ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

D2116-1G D2116-5G
TCD2116-001GEA 370 USD InStock
TCD2116-001G TCD2116-5G
3,6-Diaminocarbazole ≥98.0% (by titrimetric analysis)
3,6-Diaminocarbazole
CAS Number: 86-71-5
MDL Number: MFCD00143081
Molecular Formula: C12H11N3
Molecular Weight: 197.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 9H-Carbazole-3,6-diamine


Formula: C₁₂H₁₁N₃
MDL Number: MFCD00143081
CAS Number: 86-71-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d608cd96dde4e7b0172aab7c2a34944965cf643&vid=3d608cd96dde4e7b0172aab7c2a34944965cf643&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty