Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dioctadecylmethylamine ≥97.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

D2229-25G D2229-5G
TCD2229-25GEA 429 USD InStock
TCD2229-25G TCD2229-005G
N,N-Dioctadecylmethylamine ≥97.0% (by titrimetric analysis)
N,N-Dioctadecylmethylamine
CAS Number: 4088-22-6
MDL Number: MFCD00075495
Molecular Formula: C37H77N
Molecular Weight: 536.03
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 48

Other Product Names / Synonyms: N-Methyldioctadecylamine


Formula: C₃₇H₇₇N
Melting Pt: 48 °C
MDL Number: MFCD00075495
CAS Number: 4088-22-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2b5b557bf3310ea5c29e00bd15de3eba9e2eaa3&vid=e2b5b557bf3310ea5c29e00bd15de3eba9e2eaa3&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty