Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide ≥95.0%

Supplier: Decon Laboratories Ltd

D2259-1G D2259-100MG
TCD2259-1GEA 209.57 USD InStock
TCD2259-1G TCD2259-100MG
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide ≥95.0%
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide
[HPLC Labeling Reagent for e.e. Determination]
CAS Number: 95713-52-3
MDL Number: MFCD00042049
Molecular Formula: C9H9FN4O5
Molecular Weight: 272.19
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 229
Specific rotation [a]20/D: 55 deg (C=1, Acetone)

Product Categories: Analytical Science, e.e. Determination (HPLC Labeling Reagents), UV Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: Nalpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, Marfey's Reagent


Formula: C₉H₉FN₄O₅
MW: 272.192 g/mol
Melting Pt: 229 °C
MDL Number: MFCD00042049
CAS Number: 95713-52-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c76dd899ef2dbe4a1e5505ffbe5fbdf5c1c404c5&vid=c76dd899ef2dbe4a1e5505ffbe5fbdf5c1c404c5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty