Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-2,3-Dichloro-1,4-dioxane ≥96.0%

Supplier: Decon Laboratories Ltd

D2279-25G D2279-5G
TCD2279-025GEA 212.4 USD InStock
TCD2279-025G TCD2279-005G
trans-2,3-Dichloro-1,4-dioxane ≥96.0%
trans-2,3-Dichloro-1,4-dioxane
CAS Number: 3883-43-0
MDL Number: MFCD00191421
Molecular Formula: C4H6Cl2O2
Molecular Weight: 156.99
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 82
Freezing point (°C): 27

Other Product Names / Synonyms: trans-2,3-Dichloro-1,4-diethylene Dioxide


Formula: C₄H₆Cl₂O₂
MW: 156.99 g/mol
Boiling Pt: 82 °C
MDL Number: MFCD00191421
CAS Number: 3883-43-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5e6f5393d2cb93ba664fbb2f1b0f5575d913d15&vid=a5e6f5393d2cb93ba664fbb2f1b0f5575d913d15&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty