Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dicyano-5-methylpyrazine ≥99.0%

Supplier: Decon Laboratories Ltd

D2281-25G D2281-5G
TCD2281-025GEA 668 USD InStock
TCD2281-025G TCD2281-5G
2,3-Dicyano-5-methylpyrazine ≥99.0%
2,3-Dicyano-5-methylpyrazine
CAS Number: 52197-12-3
MDL Number: MFCD00191635
Molecular Formula: C7H4N4
Molecular Weight: 144.14
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 100

Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 5-Methyl-2,3-pyrazinedicarbonitrile


Formula: C₇H₄N₄
Melting Pt: 100 °C
MDL Number: MFCD00191635
CAS Number: 52197-12-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5137623cdaf33dc311187493d9be120651710289&vid=5137623cdaf33dc311187493d9be120651710289&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >99.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty