Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Dioxene ≥98.0%

Supplier: Decon Laboratories Ltd

D2287-1G D2287-5G D2287-25G
TCD2287-001GEA 202 USD InStock
TCD2287-001G TCD2287-005G TCD2287-25G
1,4-Dioxene ≥98.0%
1,4-Dioxene
CAS Number: 543-75-9
MDL Number: MFCD00082537
Molecular Formula: C4H6O2
Molecular Weight: 86.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 94
Flash Point (°C): 1
Specific Gravity (20/20): 1.08

Other Product Names / Synonyms: 2,3-Dihydro-1,4-dioxine


Formula: C₄H₆O₂
MDL Number: MFCD00082537
CAS Number: 543-75-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8e6e352e3facd546febdc796bfa9c70ed10ed27&vid=b8e6e352e3facd546febdc796bfa9c70ed10ed27&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty