Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Dimethyldecylamine ≥95.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

D2288-25ML D2288-500ML
TCD2288-025MLEA 21.38 USD InStock
TCD2288-025ML TCD2288-500ML
N,N-Dimethyldecylamine ≥95.0% (by GC, titration analysis)
N,N-Dimethyldecylamine
CAS Number: 1120-24-7
MDL Number: MFCD00015060
Molecular Formula: C12H27N
Molecular Weight: 185.36
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 235
Flash Point (°C): 92
Specific Gravity (20/20): 0.78

Other Product Names / Synonyms: 1-(Dimethylamino)decane


Formula: C₁₂H₂₇N
Boiling Pt: 235 °C
Density: 0.78 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00015060
CAS Number: 1120-24-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd3d08d1517ed41ccfa2d9b0ceb737c1d1214f87&vid=dd3d08d1517ed41ccfa2d9b0ceb737c1d1214f87&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (GC,T)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty