Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dichloro-2-fluorobenzene ≥98.0% for synthesis

Supplier: Decon Laboratories Ltd
Warning

D2485-25G D2485-5G
TCD2485-025GEA 38 USD InStock
TCD2485-025G TCD2485-5G
1,3-Dichloro-2-fluorobenzene ≥98.0% for synthesis
1,3-Dichloro-2-fluorobenzene
CAS Number: 2268-05-5
MDL Number: MFCD00010307
Molecular Formula: C6H3Cl2F
Molecular Weight: 164.99
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 171
Melting point (°C): 39
Flash Point (°C): 60
Storage Temperature: 0-10°C

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1,3-Dichloro-2-fluorobenzene


Formula: C₆H₃Cl₂F
MW: 164.99 g/mol
Boiling Pt: 168
Melting Pt: 39
Flash Pt: 60 °C
MDL Number: MFCD00010307
CAS Number: 2268-05-5
UN: 1325
ADR: 4.1 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c1f375c74264fdaac9dacad422b0335d461ac21&vid=2c1f375c74264fdaac9dacad422b0335d461ac21&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty