Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dodecanedioyl dichloride ≥98.0%

Supplier: Decon Laboratories Ltd

D2544-5G D2544-25G
TCD2544-005GEA 58 USD InStock
TCD2544-005G TCD2544-025G
Dodecanedioyl dichloride ≥98.0%
Dodecanedioyl dichloride
CAS Number: 4834-98-4 MDL Number: MFCD00012243 Molecular Formula: C12H20Cl2O2 Molecular Weight: 267.19 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Flash Point (°C): 110 Specific Gravity (20/20): 1.08


Formula: C₁₂H₂₀Cl₂O₂
MW: 267.19 g/mol
Density: 1.08 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00012243
CAS Number: 4834-98-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ea93a255da24a43eeefa4344e40bf940477a07a&vid=7ea93a255da24a43eeefa4344e40bf940477a07a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty