Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3-Diphenylpropionitrile ≥97.0%

Supplier: Decon Laboratories Ltd

D2590-25G D2590-5G
TCD2590-025GEA 221.8 USD InStock
TCD2590-025G TCD2590-005G
3,3-Diphenylpropionitrile ≥97.0%
3,3-Diphenylpropionitrile
CAS Number: 2286-54-6
MDL Number: MFCD00129747
Molecular Formula: C15H13N
Molecular Weight: 207.28
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 174
Melting point (°C): 86

Other Product Names / Synonyms: 2-Cyano-1,1-diphenylethane


Formula: C₁₅H₁₃N
Boiling Pt: 174 °C
Melting Pt: 86 °C
MDL Number: MFCD00129747
CAS Number: 2286-54-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=174d4aa51ef147c6c70f1db48ce8f22373f4631b&vid=174d4aa51ef147c6c70f1db48ce8f22373f4631b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty