Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dimethyl-4-nitrophenol ≥98.0%

Supplier: Decon Laboratories Ltd

D2608-25G D2608-5G
TCD2608-25GEA 357 USD InStock
TCD2608-25G TCD2608-005G
2,3-Dimethyl-4-nitrophenol ≥98.0%
2,3-Dimethyl-4-nitrophenol
CAS Number: 19499-93-5
MDL Number: MFCD01861284
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 125

Other Product Names / Synonyms: 4-Nitro-2,3-xylenol


Formula: C₈H₉NO₃
Melting Pt: 125 °C
MDL Number: MFCD01861284
CAS Number: 19499-93-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c26b9ed79e244f23af59ada4c597957474e7b496&vid=c26b9ed79e244f23af59ada4c597957474e7b496&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty