Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichlorodihexylsilane ≥90.0%

Supplier: Decon Laboratories Ltd

D2712-5G D2712-25G
TCD2712-005GEA 160 USD InStock
TCD2712-005G TCD2712-025G
Dichlorodihexylsilane ≥90.0%
Dichlorodihexylsilane
CAS Number: 18204-93-8
MDL Number: MFCD00053203
Molecular Formula: C12H26Cl2Si
Molecular Weight: 269.33
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 195
Flash Point (°C): 88
Specific Gravity (20/20): 0.97


Formula: C₁₂H₂₆Cl₂Si
Boiling Pt: 195 °C
Density: 0.97 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00053203
CAS Number: 18204-93-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27b280dc3e5f2fd26663ce2975cb56c5ba9d45a4&vid=27b280dc3e5f2fd26663ce2975cb56c5ba9d45a4&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >90.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty