Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethylmalonyl dichloride ≥98.0%

Supplier: Decon Laboratories Ltd

D2723-5G D2723-25G
TCD2723-005GEA 151.11 USD InStock
TCD2723-005G TCD2723-25G
Dimethylmalonyl dichloride ≥98.0%
Dimethylmalonyl dichloride
CAS Number: 5659-93-8
MDL Number: MFCD00192081
Molecular Formula: C5H6Cl2O2
Molecular Weight: 169.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 51
Specific Gravity (20/20): 1.27


Formula: C₅H₆Cl₂O₂
MW: 169.00 g/mol
Density: 1.27 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00192081
CAS Number: 5659-93-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=742c275f5ccee4e7910379b0db2dbe2f6aca746a&vid=742c275f5ccee4e7910379b0db2dbe2f6aca746a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty