D3305-25G
D3305-5G
TCD3305-25GEA
112
USD
InStock
TCD3305-25G
TCD3305-5G
Dimethyl-DL-glutamate hydrochloride ≥98.0%
Dimethyl-DL-glutamate hydrochloride
CAS Number: 13515-99-6
Molecular Formula: C7H13NO4
Molecular Weight: 211.64
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 154
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: DL-Glutamic Acid Dimethyl Ester Hydrochloride, H-DL-Glu(OMe)-OMe.HCl