Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diisopropyl allylboronate ≥98.0%

Supplier: Decon Laboratories Ltd

D3738-5G D3738-1G
TCD3738-5GEA 687 USD InStock
TCD3738-5G TCD3738-1G
Diisopropyl allylboronate ≥98.0%
Diisopropyl allylboronate
CAS Number: 51851-79-7
Molecular Formula: C9H19BO2
Molecular Weight: 170.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Storage Temperature: 0-10°C

Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: Allylboronic Acid Diisopropyl Ester


Formula: C₉H₁₉BO₂
MW: 170.06 g/mol
CAS Number: 51851-79-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec68e231a5980d6ce28a17623380aa09ac444542&vid=ec68e231a5980d6ce28a17623380aa09ac444542&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty