Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Desloratadine ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd

D3787-100MG D3787-1G
TCD3787-100MGEA 35 USD InStock
TCD3787-100MG TCD3787-1G
Desloratadine ≥98.0% (by HPLC, titration analysis)
Desloratadine
CAS Number: 100643-71-8
MDL Number: MFCD00871949
Molecular Formula: C19H19ClN2
Molecular Weight: 310.83
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 157
Storage Temperature: 0-10°C


Formula: C₁₉H₁₉ClN₂
Melting Pt: 157 °C
MDL Number: MFCD00871949
CAS Number: 100643-71-8
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8298d613288a0f59cb4d5cce127cc073b369fc63&vid=8298d613288a0f59cb4d5cce127cc073b369fc63&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty