Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1'-Difluoro-2,2'-bipyridinium bis(tetrafluoroborate) ≥95.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

D3812-1G D3812-5G
TCD3812-1GEA 74 USD InStock
TCD3812-1G TCD3812-5G
1,1'-Difluoro-2,2'-bipyridinium bis(tetrafluoroborate) ≥95.0% (by titrimetric analysis)
1,1'-Difluoro-2,2'-bipyridinium bis(tetrafluoroborate)
CAS Number: 178439-26-4
MDL Number: MFCD01073546
Molecular Formula: C10H8B2F10N2
Molecular Weight: 367.79
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Storage Temperature: 0-10°C


Formula: C₁₀H₈B₂F₁₀N₂
MDL Number: MFCD01073546
CAS Number: 178439-26-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a773816a66c4c5fae2f6f1ba0ab5dcf582adacad&vid=a773816a66c4c5fae2f6f1ba0ab5dcf582adacad&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty