Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dibromo-1H-pyrrole-2,5-dione ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd

D3910-1G D3910-5G
TCD3910-1GEA 75 USD InStock
TCD3910-1G TCD3910-5G
3,4-Dibromo-1H-pyrrole-2,5-dione ≥98.0% (by HPLC, titration analysis)
3,4-Dibromo-1H-pyrrole-2,5-dione
CAS Number: 1122-10-7
MDL Number: MFCD00185696
Molecular Formula: C4HBr2NO2
Molecular Weight: 254.87
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 228
Lambda max.: 243 nm (MeOH)


Formula: C4HBr2NO2
Melting Pt: 228 °C
MDL Number: MFCD00185696
CAS Number: 1122-10-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad219aee3053940fd20f2ea5781d79a9a6098249&vid=ad219aee3053940fd20f2ea5781d79a9a6098249&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty