Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-4-ethylbenzenesulfonate ≥98.0%

Supplier: Decon Laboratories Ltd

E0104-25G E0104-500G
TCE0104-025GEA 75 USD InStock
TCE0104-025G TCE0104-500G
Sodium-4-ethylbenzenesulfonate ≥98.0%
Sodium-4-ethylbenzenesulfonate
CAS Number: 14995-38-1
MDL Number: MFCD00070568
Molecular Formula: C8H10O3S
Molecular Weight: 208.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal

Other Product Names / Synonyms: 4-Ethylbenzenesulfonic Acid Sodium Salt


Formula: C₈H₁₀O₃S
MDL Number: MFCD00070568
CAS Number: 14995-38-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcffe4c20d5496bba52a5846a88e011d237c187c&vid=bcffe4c20d5496bba52a5846a88e011d237c187c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty