Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Acetyl-5-hydroxytryptamine ≥98.0% (by HPLC, total nitrogen)

Supplier: Decon Laboratories Ltd

A1277-1G A1277-100MG
TCA1277-001GEA 454 USD InStock
TCA1277-001G TCA1277-100MG
N-Acetyl-5-hydroxytryptamine ≥98.0% (by HPLC, total nitrogen)
N-Acetyl-5-hydroxytryptamine
CAS Number: 1210-83-9
MDL Number: MFCD00005656
Molecular Formula: C12H14N2O2
Molecular Weight: 218.26
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 120

Other Product Names / Synonyms: 3-(2-Acetamidoethyl)-5-hydroxyindole, N-Acetylserotonin


Formula: C₁₂H₁₄N₂O₂
Melting Pt: 120 °C
MDL Number: MFCD00005656
CAS Number: 1210-83-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89784f3057acb74da7892bb4fbbc772d39ff813b&vid=89784f3057acb74da7892bb4fbbc772d39ff813b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,N)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty