Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethylpyrrolidine ≥98.0%

Supplier: Decon Laboratories Ltd

E0510-25G E0510-5G
TCE0510-025GEA 196.31 USD InStock
TCE0510-025G TCE0510-5G
1-Ethylpyrrolidine ≥98.0%
1-Ethylpyrrolidine
CAS Number: 7335-06-0
MDL Number: MFCD01632186
Molecular Formula: C6H13N
Molecular Weight: 99.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Flash Point (°C): -22
Specific Gravity (20/20): 0.81


Formula: C₆H₁₃N
Boiling Pt: 105 °C
Density: 0.81 (20 °C)
Flash Pt: –22 °C
MDL Number: MFCD01632186
CAS Number: 7335-06-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f3c9ba619b30ba4d09b0d72fee4c54424495112&vid=6f3c9ba619b30ba4d09b0d72fee4c54424495112&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty