Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Epinastine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: Decon Laboratories Ltd

E0799-100MG E0799-1G
TCE0799-100MGEA 66 USD InStock
TCE0799-100MG TCE0799-1G
Epinastine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Epinastine hydrochloride
CAS Number: 108929-04-0
MDL Number: MFCD00933434
Molecular Formula: C16H15N3
Molecular Weight: 285.78
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 275
Storage Temperature: 0-10°C

Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents
Other Product Names / Synonyms: 3-Amino-9,13b-dihydro-1H-dibenzo[c,f]imidazo[1,5-a]azepine Hydrochloride


Formula: C₁₆H₁₅N₃
MDL Number: MFCD00933434
CAS Number: 108929-04-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c12c5d20e0d758dca8e861519ec332ff53d2fa21&vid=c12c5d20e0d758dca8e861519ec332ff53d2fa21&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty