Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-(2,6-dichloro-5-fluoro-3-pyridyl)-3-oxopropionate ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

E0811-25G E0811-5G
TCE0811-25GEA 177 USD InStock
TCE0811-25G TCE0811-5G
Ethyl-3-(2,6-dichloro-5-fluoro-3-pyridyl)-3-oxopropionate ≥98.0% (by titrimetric analysis)
Ethyl-3-(2,6-dichloro-5-fluoro-3-pyridyl)-3-oxopropionate
CAS Number: 96568-04-6
MDL Number: MFCD00799535
Molecular Formula: C10H8Cl2FNO3
Molecular Weight: 280.08
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 69

Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction), Fluorinated Building Blocks
Other Product Names / Synonyms: 3-(2,6-Dichloro-5-fluoro-3-pyridyl)-3-oxopropionic Acid Ethyl Ester, Ethyl (2,6-Dichloro-5-fluoronicotinoyl)acetate, (2,6-Dichloro-5-fluoronicotinoyl)acetic Acid Ethyl Ester


Formula: C₁₀H₈Cl₂FNO₃
Melting Pt: 69 °C
MDL Number: MFCD00799535
CAS Number: 96568-04-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=343fdb51f0ac06dfc98382ac3b83f5882d542e5a&vid=343fdb51f0ac06dfc98382ac3b83f5882d542e5a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty