Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluorofluorene ≥98.0%

Supplier: Decon Laboratories Ltd

F0228-5G F0228-25G
TCF0228-005GEA 174.6 USD InStock
TCF0228-005G TCF0228-025G
2-Fluorofluorene ≥98.0%
2-Fluorofluorene
CAS Number: 343-43-1
MDL Number: MFCD00031477
Molecular Formula: C13H9F
Molecular Weight: 184.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 98


Formula: C₁₃H₉F
Melting Pt: 98 °C
MDL Number: MFCD00031477
CAS Number: 343-43-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4aa632d5c183878212bf7d66bcea7ff85f05f453&vid=4aa632d5c183878212bf7d66bcea7ff85f05f453&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty