Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Famotidine ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

F0530-25G F0530-5G
TCF0530-25GEA 348 USD InStock
TCF0530-25G TCF0530-5G
Famotidine ≥98.0% (by titrimetric analysis)
Famotidine
CAS Number: 76824-35-6
Molecular Formula: C8H15N7O2S3
Molecular Weight: 337.44
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 160

Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 3-(2-Guanidinothiazol-4-ylmethylthio)-N1-sulfamoylpropionamide


Formula: C₈H₁₅N₇O₂S₃
Melting Pt: 160 °C
MDL Number: MFCD00079297
CAS Number: 76824-35-6
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96487b98e8a04fee2db1283a269e69e0decc4346&vid=96487b98e8a04fee2db1283a269e69e0decc4346&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty