Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-Glu-OH ≥97.0%

Supplier: Decon Laboratories Ltd

F0600-25G F0600-5G
TCF0600-25GEA 165.3 USD InStock
TCF0600-25G TCF0600-5G
Fmoc-D-Glu-OH ≥97.0%
Fmoc-D-Glu-OH
CAS Number: 104091-09-0
Molecular Formula: C20H19NO6
Molecular Weight: 369.37
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: 19 deg (C=1, DMF)

Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-D-glutamic Acid, Fmoc-D-Glu-OH


Formula: C₂₀H₁₉NO₆
MDL Number: MFCD09037362
CAS Number: 104091-09-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e353ccbdbb8a1500d62abef1c0e6a68129f54989&vid=e353ccbdbb8a1500d62abef1c0e6a68129f54989&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty