Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Flavoxate hydrochloride ≥98.0%

Supplier: Decon Laboratories Ltd

F0717-25G F0717-5G
TCF0717-25GEA 214.5 USD InStock
TCF0717-25G TCF0717-5G
Flavoxate hydrochloride ≥98.0%
Flavoxate hydrochloride
CAS Number: 3717-88-2 MDL Number: MFCD00072099 Molecular Formula: C24H25NO4 Molecular Weight: 427.93 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 234


Formula: C₂₄H₂₅NO₄
MW: 427.93 g/mol
Melting Pt: 234 °C
MDL Number: MFCD00072099
CAS Number: 3717-88-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=104ab60e0a508e8baf527ad08381fdf39446803c&vid=104ab60e0a508e8baf527ad08381fdf39446803c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty