Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Guanidine sulfamate ≥90.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

G0022-25G G0022-500G
TCG0022-025GEA 29 USD InStock
TCG0022-025G TCG0022-500G
Guanidine sulfamate ≥90.0% (by titrimetric analysis)
Guanidine sulfamate
CAS Number: 50979-18-5 MDL Number: MFCD00060261 Molecular Formula: CH5N3 Molecular Weight: 156.16 Purity/Analysis Method: 90.0% (T) Form: Crystal


Formula: CH₅N₃
MDL Number: MFCD00060261
CAS Number: 50979-18-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53e0e5cb249a426b655bef8f5d6b2701b81953af&vid=53e0e5cb249a426b655bef8f5d6b2701b81953af&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 90.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty