Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diguanidine phosphate ≥95.0%

Supplier: Decon Laboratories Ltd

G0165-500G G0165-25G
TCG0165-500GEA 113.93 USD InStock
TCG0165-500G TCG0165-025G
Diguanidine phosphate ≥95.0%
Diguanidine phosphate
CAS Number: 5423-23-4
MDL Number: MFCD00039131
Molecular Formula: CH5N3
Molecular Weight: 216.14
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Formula: C₂H₁₃N₆O₄P
MW: 216.14 g/mol
MDL Number: MFCD00039131
CAS Number: 5423-23-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c92e28968da9a39da338dee2edbe7a0646b28b2a&vid=c92e28968da9a39da338dee2edbe7a0646b28b2a&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >95.0% (T)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty