Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Gulose ≥98.0% (by HPLC)

Supplier: Decon Laboratories Ltd

G0239-25G G0239-1G G0239-5G
TCG0239-25GEA 957 USD InStock
TCG0239-25G TCG0239-001G TCG0239-005G
L-Gulose ≥98.0% (by HPLC)
L-Gulose
CAS Number: 6027-89-0 MDL Number: MFCD00136022 Molecular Formula: C6H12O6 Molecular Weight: 180.16 Purity/Analysis Method: 98.0% (HPLC) Form: Crystal Melting point (°C): 132 Specific rotation [a]20/D: 25 deg (C=0.5, H2O)


Formula: C₆H₁₂O₆
Melting Pt: 132 °C
MDL Number: MFCD00136022
CAS Number: 6027-89-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6845218e289bec51651889852b9bcaebbf58ead4&vid=6845218e289bec51651889852b9bcaebbf58ead4&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty