Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Gabapentin ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

G0318-5G G0318-25G
TCG0318-5GEA 75.3 USD InStock
TCG0318-5G TCG0318-25G
Gabapentin ≥98.0% (by GC, titration analysis)
Gabapentin
CAS Number: 60142-96-3
MDL Number: MFCD00865286
Molecular Formula: C9H17NO2
Molecular Weight: 171.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 166

Product Categories: Bioscience, Neurotransmitter Agent Ingredients, Voltage-dependent Calcium Channel (VDCC) Antagonists
Other Product Names / Synonyms: 1-(Aminomethyl)cyclohexaneacetic Acid


Formula: C₉H₁₇NO₂
Melting Pt: 166 °C
MDL Number: MFCD00865286
CAS Number: 60142-96-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3acfeae76416b20889ef291232214a218afdaaa4&vid=3acfeae76416b20889ef291232214a218afdaaa4&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty