Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Gliquidone ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd

G0332-5G G0332-1G
TCG0332-5GEA 167 USD InStock
TCG0332-5G TCG0332-1G
Gliquidone ≥98.0% (by HPLC, titration analysis)
Gliquidone
CAS Number: 33342-05-1 MDL Number: MFCD00631870 Molecular Formula: C27H33N3O6S Molecular Weight: 527.64 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 179


Formula: C₂₇H₃₃N₃O₆S
Melting Pt: 179 °C
MDL Number: MFCD00631870
CAS Number: 33342-05-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1227c6a55161c4a05937cbb64ef4d6bde9ce303&vid=c1227c6a55161c4a05937cbb64ef4d6bde9ce303&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty