Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitrophenyl hexanoate ≥98.0%

Supplier: Decon Laboratories Ltd

H0484-1G H0484-5G
TCH0484-001GEA 93 USD InStock
TCH0484-001G TCH0484-005G
4-Nitrophenyl hexanoate ≥98.0%
4-Nitrophenyl hexanoate
CAS Number: 956-75-2
MDL Number: MFCD00056111
Molecular Formula: C12H15NO4
Molecular Weight: 237.26
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Specific Gravity (20/20): 1.14
Storage Temperature: 0-10°C

Other Product Names / Synonyms: Hexanoic Acid 4-Nitrophenyl Ester


Formula: C₁₂H₁₅NO₄
Boiling Pt: 145 °C
Density: 1.14 (20 °C)
MDL Number: MFCD00056111
CAS Number: 956-75-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2b07ee043cec728e9143c0115e3d878198b7954&vid=e2b07ee043cec728e9143c0115e3d878198b7954&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty