Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexamethyleneammonium hexamethylenedithiocarbamate ≥97.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

H0519-5G H0519-25G
TCH0519-005GEA 35 USD InStock
TCH0519-005G TCH0519-025G
Hexamethyleneammonium hexamethylenedithiocarbamate ≥97.0% (by titrimetric analysis)
Hexamethyleneammonium hexamethylenedithiocarbamate
CAS Number: 2608-11-9
MDL Number: MFCD00036327
Molecular Formula: C6H13N
Molecular Weight: 274.49
Purity/Analysis Method: >97.0% (T)
Form: Crystal

Other Product Names / Synonyms: Hexamethylenedithiocarbamic Acid Hexamethyleneammonium Salt


Formula: C₆H₁₃N
MDL Number: MFCD00036327
CAS Number: 2608-11-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ec105eb8fdafbbc70b62da1946c22cc9e594792&vid=0ec105eb8fdafbbc70b62da1946c22cc9e594792&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >97.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty