Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

m-Anisohydrazide ≥98.0% (by GC, titration analysis)

Supplier: Decon Laboratories Ltd

A1446-5G A1446-25G
TCA1446-005GEA 72 USD InStock
TCA1446-005G TCA1446-025G
m-Anisohydrazide ≥98.0% (by GC, titration analysis)
m-Anisohydrazide
CAS Number: 5785-06-8
MDL Number: MFCD00007601
Molecular Formula: C8H10N2O2
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 93

Other Product Names / Synonyms: m-Anisic Hydrazide, m-Anisoylhydrazine, 3-Methoxybenzoylhydrazine


Formula: C₈H₁₀N₂O₂
Melting Pt: 93 °C
MDL Number: MFCD00007601
CAS Number: 5785-06-8
EINECS: 227-310-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19b45ba9aeb3ed95ca28c67bbe092935fb9a6ced&vid=19b45ba9aeb3ed95ca28c67bbe092935fb9a6ced&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty