Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[4-(Hydroxymethyl)-2-nitrophenoxy]ethanol ≥98.0%

Supplier: Decon Laboratories Ltd

H1048-1G H1048-100MG
TCH1048-1GEA 321.2 USD InStock
TCH1048-1G TCH1048-100MG
2-[4-(Hydroxymethyl)-2-nitrophenoxy]ethanol ≥98.0%
2-[4-(Hydroxymethyl)-2-nitrophenoxy]ethanol
MDL Number: MFCD04117919
Molecular Formula: C9H11NO5
Molecular Weight: 213.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal

Other Product Names / Synonyms: Ethylene Glycol Mono-4-(hydroxymethyl)-2-nitrophenyl Ether, 4-(2-Hydroxyethoxy)-3-nitrobenzyl Alcohol


https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=426429313980c6aa387e822f0d249e020042dbfd&vid=426429313980c6aa387e822f0d249e020042dbfd&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty