Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Harmaline ≥98.0% (by HPLC, titration analysis)

Supplier: Decon Laboratories Ltd

H1237-1G H1237-5G
TCH1237-1GEA 83 USD InStock
TCH1237-1G TCH1237-5G
Harmaline ≥98.0% (by HPLC, titration analysis)
Harmaline
CAS Number: 304-21-2
MDL Number: MFCD00004955
Molecular Formula: C13H14N2O
Molecular Weight: 214.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 251

Product Categories: Bioscience, Indole Alkaloids
Other Product Names / Synonyms: 3,4-Dihydro-7-methoxy-1-methyl-beta-carboline, 3,4-Dihydro-7-methoxy-1-methyl-9H-pyrido[3,4-b]indole, Dihydroharmine


Formula: C₁₃H₁₄N₂O
Melting Pt: 251 °C
MDL Number: MFCD00004955
CAS Number: 304-21-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ed73f20deb1dd862e4caeec5a24fa952517ffc3&vid=2ed73f20deb1dd862e4caeec5a24fa952517ffc3&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (HPLC,T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty