Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethoxyethyl isobutyrate ≥98.0%

Supplier: Decon Laboratories Ltd

I0282-5ML I0282-25ML
TCI0282-005MLEA 28 USD InStock
TCI0282-005ML TCI0282-025ML
2-Ethoxyethyl isobutyrate ≥98.0%
2-Ethoxyethyl isobutyrate
CAS Number: 54396-97-3
MDL Number: MFCD00009250
Molecular Formula: C8H16O3
Molecular Weight: 160.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 57
Specific Gravity (20/20): 0.93

Other Product Names / Synonyms: Cellosolve Isobutyrate, Isobutyric Acid 2-Ethoxyethyl Ester, Ethylene Glycol Monoethyl Ether Isobutyrate


Formula: C₈H₁₆O₃
MW: 160.21 g/mol
Boiling Pt: 85 °C
Density: 0.93 (20 °C)
Flash Pt: 57 °C
MDL Number: MFCD00009250
CAS Number: 54396-97-3
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de298302afde874dc18524b88244c7b9185827be&vid=de298302afde874dc18524b88244c7b9185827be&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (GC)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty