Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Isovalerylglycine ≥98.0% (by titrimetric analysis)

Supplier: Decon Laboratories Ltd

I0301-1G I0301-10G
TCI0301-001GEA 120.7 USD InStock
TCI0301-001G TCI0301-010G
N-Isovalerylglycine ≥98.0% (by titrimetric analysis)
N-Isovalerylglycine
CAS Number: 16284-60-9
MDL Number: MFCD00059629
Molecular Formula: C7H13NO3
Molecular Weight: 159.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 105

Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Isovaleroylglycine


Formula: C₇H₁₃NO₃
Melting Pt: 105 °C
MDL Number: MFCD00059629
CAS Number: 16284-60-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48f6e54fc6daa50c7ccb9e7fb32bea45180df763&vid=48f6e54fc6daa50c7ccb9e7fb32bea45180df763&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Purity/Analysis Method >98.0% (T)

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty